Please login to your online account to display your discounted pricing
Amniomax™ C-100 Supplement DFS Item
A part of kit used to prepare fully supplemented Gibco AmnioMAX C-100 Complete Medium developed for short term culture of human amniotic fluid cells for cytogenetic studies and in vitro diagnostic procedures
Manufacturer: life technologies  LS12556015
 View more versions of this product
 Disclaimers
In vitro diagnostic
Catalog No. 12556015
  • / Each
Description & Specifications
Specifications
Quantity 15mL
Storage Requirements Storage: -5 to -20°C. Shelf life: 16 months from date of manufacture. Use complete medium within 7 to 10 days once opened.
Packaging Sustainable packaging