missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ 6Ckine Recombinant Human Protein DFS Item

Recombinant 6Ckine is a bioactive protein intended for use in cell culture applications

$171.15 - $171.15

Specifications

Purity 95%
Purification Method Ion Exchange
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias 6Ckine
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PHC1474 Gibco™
LSPHC1474
10μg Each for $171.15
Specifications

Description

Recombinant human 6Ckine is intended for use in cell culture applications. 6Ckine is a chemokine involved in chemotaxis of specific cell types.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

95%
<0.1ng/μg
6Ckine
Ion Exchange
Lyophilized
6366