Please login to your online account to display your discounted pricing

Gibco™ 6Ckine Recombinant Human Protein DFS Item


Recombinant 6Ckine is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHC1474

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PHC1474

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at 2° to 8°C.
Source Human
Purification Method Ion Exchange
Purity 95%
Quantity 10μg
Molecular Weight 12kDa
Endotoxin Concentration <0.1ng/μg
Validated Application Cell Culture
Expression System E. coli
Protein Subtype 6 CKINE
Protein Family Chemokines & Receptors
Structural Form Recombinant/Ligand
Gene Alias 6Ckine
Gene ID (Entrez) 6366
For Use With (Application) Cell Culture; Mammalian Cell Culture
Without Additives Carrier Protein
Format Lyophilized

Recombinant human 6Ckine is intended for use in cell culture applications. 6Ckine is a chemokine involved in chemotaxis of specific cell types.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.