Please login to your online account to display your discounted pricing

Gibco™ 6Ckine Recombinant Human Protein DFS Item


Recombinant 6Ckine is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHC1471

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PHC1471

  • / Each
 

Description & Specifications

Specifications

Protein Subtype 6 CKINE
Gene Alias 6Ckine
Expression System E. coli
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Purity 95%
Purification Method Ion Exchange
Source Human
Storage Requirements Store at 2° to 8°C.
Structural Form Recombinant/Ligand
Protein Family Chemokines & Receptors
Gene ID (Entrez) 6366
Without Additives Carrier Protein
Quantity 100μg
Molecular Weight 12kDa
For Use With (Application) Cell Culture; Mammalian Cell Culture
Validated Application Cell Culture

Recombinant human 6Ckine is intended for use in cell culture applications. 6Ckine is a chemokine involved in chemotaxis of specific cell types.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.