Please login to your online account to display your discounted pricing

Gibberellic acid potassium salt, 75%, MP Biomedicals™


Plant growth hormone

Quantity 1g

Manufacturer: mp biomedicals  10028801

 View more versions of this product

Catalog No. ICN10028801

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 384.5
Color White
Quantity 1g
Name Note Potassium salt
Chemical Name or Material Gibberellic acid
Assay Percent Range 75%
Molecular Formula C19H21O6K
CAS 125-67-7
Recommended Storage Store at 2 to 8°C

Specifications

Chemical Name or Material Gibberellic acid
Name Note Potassium salt
CAS 125-67-7
Physical Form Powder
Assay Percent Range 75%
Molecular Formula C19H21O6K
Formula Weight 384.5
Accessories

Store at 2 to 8°C