Please login to your online account to display your discounted pricing

Gibberellic acid potassium salt, 75%, MP Biomedicals™

Plant growth hormone

$136.52 - $466.30

Chemical Identifiers

Chemical Name or Material Gibberellic acid
Name Note Potassium salt
CAS 125-67-7
Physical Form Powder
Assay Percent Range 75%
Molecular Formula C19H21O6K
Formula Weight 384.5
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10028801

 
mp biomedicals
10028801
1g Each for $136.52

ICN10028805

 
mp biomedicals
10028805
5g Each for $466.30
Description & Specifications

Specifications

Formula Weight 384.5
Physical Form Powder
Color White
Quantity 1g
Name Note Potassium salt
Assay Percent Range 75%
Recommended Storage Store at 2 to 8°C
Molecular Formula C19H21O6K
CAS 125-67-7
Chemical Name or Material Gibberellic acid
Safety and Handling

Store at 2 to 8°C