Please login to your online account to display your discounted pricing

Gibberellic Acid, Potassium Salt 10%, MP Biomedicals™


Gibberellic acid is an endogenous plant growth regulator

Quantity 5g

Manufacturer: mp biomedicals  10028205

 View more versions of this product

Catalog No. ICN10028205

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 384.5
Quantity 5g
EINECS Number 204-749-6
Physical State Solid
Chemical Name or Material Gibberellic Acid, Potassium Salt
Molecular Formula C19H21O6.K
MDL Number MFCD00083342
Synonym GA3
Assay Percent Range 10%
CAS 125-67-7
Recommended Storage Store at 2° to 8°C.

Specifications

Chemical Name or Material Gibberellic Acid, Potassium Salt
Assay Percent Range 10%
Molecular Formula C19H21O6.K
Formula Weight 384.5
MDL Number MFCD00083342
Synonym GA3
Accessories

Store at 2° to 8°C.