missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibberellic Acid, Potassium Salt 10%, MP Biomedicals™

Gibberellic acid is an endogenous plant growth regulator

$477.74 - $825.59

Chemical Identifiers

Assay Percent Range 10%
Molecular Formula C19H21O6.K
Formula Weight 384.5
MDL Number MFCD00083342
Synonym GA3
Chemical Name or Material Gibberellic Acid, Potassium Salt
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10028210 Mp Biomedicals
10028210
10g Each for $825.59
ICN10028205 Mp Biomedicals
10028205
5g Each for $477.74
Specifications Description & Specifications

Chemical Identifiers

10%
384.5
GA3
C19H21O6.K
MFCD00083342
Gibberellic Acid, Potassium Salt

Specifications

384.5
Solid
125-67-7
GA3
Gibberellic Acid, Potassium Salt
10g
MFCD00083342
C19H21O6.K
10%

Safety and Handling

EINECSNumber : 204-749-6

Recommended Storage : Store at 2° to 8°C.