Please login to your online account to display your discounted pricing

Alfa Aesar™ Gibberellic Acid A4, 90%

Chemical Identifiers

Chemical Name or Material Gibberellic Acid A4
CAS 468-44-0
Formula Weight 332.39
MDL Number MFCD00133363
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66065LB0

  SDS
alfa aesar™
J66065LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 332.39
Chemical Name or Material Gibberellic Acid A4
Assay 90%
CAS 468-44-0
Physical Form Powder
MDL Number MFCD00133363
Boiling Point 574°C
Storage Note 1 -30° to -10°C
Quantity 5mg
Color White to off-white
Solubility Information Soluble in methanol or ethanol; Also soluble in aqueous sodium bicarbonate or sodium acetate

  • Used for its stimulating effect on cell division and cell elongation, seed germination and flowering in long day plants

Safety and Handling

-30° to -10°C