Please login to your online account to display your discounted pricing

Gibberellic acid, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC410910250

 View more versions of this product

Catalog No. AC410910250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 346.38
Quantity 25g
Color White to pale yellow
Water 1% Max. (K.F.)
Specific Rotation Condition 82.50 (20.00°C c=10,ethanol)
Merck Index 15, 4454
Infrared Spectrum Authentic
Chemical Name or Material Gibberellic acid
Molecular Formula C19H22O6
Assay Percent Range 99%
CAS 77-06-5
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Warning
MDL Number MFCD00079329
Specific Rotation +71° to +81° (20°C, 589nm) (c=2, methanol)
Synonym 2,4a,7-Trihydroxy-1-methyl-8-methylenegibb-3-ene-1,10-dicarboxylic acid 1,4a-lactone
Packaging Glass bottle
Health Hazard 2 GHS P Statement
Causes serious eye irritation.

Specifications

Chemical Name or Material Gibberellic acid
CAS 77-06-5
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C19H22O6
Formula Weight 346.38
MDL Number MFCD00079329
Synonym 2,4a,7-Trihydroxy-1-methyl-8-methylenegibb-3-ene-1,10-dicarboxylic acid 1,4a-lactone
Accessories

Warning

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes serious eye irritation.

SDS & Safety Documents