missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibberellic acid, 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 410910010

 View more versions of this product

Catalog No. AC410910010


 

Specifications Description & Specifications

Specifications

≥98.5%
346.38
White to pale yellow
1% Max. (K.F.)
+71° to +81° (20°C, 589nm) (c=2, methanol)
C19H22O6
99%
2,4a,7-Trihydroxy-1-methyl-8-methylenegibb-3-ene-1,10-dicarboxylic acid 1,4a-lactone
Authentic
Crystalline Powder
1g
15, 4454
82.50 (20.00°C c=10,ethanol)
Gibberellic acid
77-06-5
MFCD00079329
Glass bottle

Chemical Identifiers

77-06-5
C19H22O6
MFCD00079329
Gibberellic acid
99%
346.38
2,4a,7-Trihydroxy-1-methyl-8-methylenegibb-3-ene-1,10-dicarboxylic acid 1,4a-lactone
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS P Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning