Please login to your online account to display your discounted pricing

Gibberellic acid, >90%, MP Biomedicals™


Quantity 1g

Manufacturer: mp biomedicals  0215118001

 View more versions of this product

Catalog No. ICN15118001

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1g
Chemical Name or Material Gibberellic acid
CAS 77-06-5
Physical Form White crystalline powder
Assay Percent Range >90%
Molecular Formula C19H22O6
Formula Weight 346.38g/mol
Synonym GA3

Specifications

Chemical Name or Material Gibberellic acid
CAS 77-06-5
Physical Form White crystalline powder
Assay Percent Range >90%
Molecular Formula C19H22O6
Formula Weight 346.38g/mol
Synonym GA3