Please login to your online account to display your discounted pricing

Gibberellic acid, >90%, MP Biomedicals™

$68.04 - $68.04

Chemical Identifiers

Chemical Name or Material Gibberellic acid
CAS 77-06-5
Physical Form White crystalline powder
Assay Percent Range >90%
Molecular Formula C19H22O6
Formula Weight 346.38g/mol
Synonym GA3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15118001

 
mp biomedicals
0215118001
1g Each for $68.04
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material Gibberellic acid
CAS 77-06-5
Physical Form White crystalline powder
Assay Percent Range >90%
Molecular Formula C19H22O6
Formula Weight 346.38g/mol
Synonym GA3