missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ GHS Labeled Right-to-Know Safety-Vented Wash Bottles

Unique valve prevents pressure build-up, eliminating potentially dangerous chemical drips and allows dispensing of wash bottle in upright or inverted position.

Manufacturer:  Fisherbrand™ 124323011

 View more versions of this product

Catalog No. 11-865-168




Description

Description

Unique valve prevents pressure build-up, eliminating potentially dangerous chemical drips and allows dispensing of wash bottle in upright or inverted position.

  • Pre-printed bottles with chemical name and formula, GHS pictogram and signal word; National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
  • Translucent LDPE bottles, Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm
  • Wide mouth 53mm (2 in.) for easy refilling
  • Color-coded polypropylene closures aid in quick and easy identification
  • Assortment bag includes one each: Acetone, Isopropanol, Bleach and Ethanol
Bottle with Label: TOLUENE
  • Red LDPE bottle
  • Does not include vented valve


Bottle with Label: DEIONIZED H2O, DISTILLED H20 and WATER
  • Does not indicate hazardous chemical and does not have/require GHS pictogram or signal word


Bottle with Label: METHYL ETHYL KETONE
  • Does not include vented valve

Compliance

Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA’s HazCom 2012 requirements – as pertains to “Workplace Labels”.

Specifications

Specifications

1000 mL
33.81 oz.
Natural
3/Bag
Green
Not Autoclavable
Polypropylene
No
Cylindrical
1000 mL
METHANOL
LDPE
Safety Vented Wash Bottle
Yes
53 mm
Cap
Wide
Yes
SDS
Documents
Product Certifications
Promotions

Promotions