missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ GHS Labeled Right-to-Know Safety-Vented Wash Bottles
Unique valve prevents pressure build-up, eliminating potentially dangerous chemical drips and allows dispensing of wash bottle in upright or inverted position.
Supplier: Fisherbrand™ 124323008
Description
Unique valve prevents pressure build-up, eliminating potentially dangerous chemical drips and allows dispensing of wash bottle in upright or inverted position.
- Pre-printed bottles with chemical name and formula, GHS pictogram and signal word; National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
- Translucent LDPE bottles, Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm
- Wide mouth 53mm (2 in.) for easy refilling
- Color-coded polypropylene closures aid in quick and easy identification
- Assortment bag includes one each: Acetone, Isopropanol, Bleach and Ethanol
Bottle with Label: TOLUENE
- Red LDPE bottle
- Does not include vented valve
Bottle with Label: DEIONIZED H2O, DISTILLED H20 and WATER
- Does not indicate hazardous chemical and does not have/require GHS pictogram or signal word
Bottle with Label: METHYL ETHYL KETONE
- Does not include vented valve
Compliance
Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHAâs HazCom 2012 requirements – as pertains to Workplace Labels.
Specifications
Polypropylene | |
Yes | |
53 mm | |
1000 mL | |
33.81 oz. | |
LDPE | |
Yellow | |
Not Autoclavable | |
Wide |
Safety Vented Wash Bottle | |
Cap | |
Isopropanol | |
1000 mL | |
Natural | |
3/Bag | |
Yes | |
No | |
Cylindrical |