missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GFP Rat, eFluor 660, Clone: 5F12.4, eBioscience™

Rat Monoclonal Antibody

$355.08 - $487.08

Specifications

Antigen GFP
Clone 5F12.4
Host Species Rat
Gene Alias Green fluorescent protein
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124419 Life Technologies
LS50649880
25μg Each for $355.08
501124420 Life Technologies
LS50649882
100μg Each for $487.08
Specifications

Description

This 5F12.4 monoclonal antibody reacts with green-fluorescent protein (GFP), which was originally isolated from the cnidarian Aequorea victoria. This protein absorbs blue light (maximally at 395 nm) and emits green light (peak at 509) without the requirement of exogenous substrates and cofactors. These unique qualities allow GFP to be used to monitor gene expression and protein localization in vivo. Several mutant forms of GFP have been developed which fluoresce more intensely and have shifted excitation maxima when compared to wildtype GFP, making them useful for flow cytometry, fluorescence microscopy, and double-labeling applications. This antibody is capable of staining formaldehyde fixed cells.
Description & Specifications

Specifications

GFP
Rat
Flow Cytometry
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
5F12.4
Green fluorescent protein
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer