Please login to your online account to display your discounted pricing

GFP Rat, Biotin, Clone: 5F12.4, eBioscience™

Rat Monoclonal Antibody

$143.55 - $433.55

Specifications

Antigen GFP
Clone 5F12.4
Host Species Rat
Gene Alias Green fluorescent protein
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50245546

 
life technologies
LS13649880
25μg Each for $143.55

50245547

 
life technologies
LS13649882
100μg Each for $433.55
Description & Specifications

Specifications

Antigen GFP
Clone 5F12.4
Host Species Rat
Gene Alias Green fluorescent protein
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

This 5F12.4 monoclonal antibody reacts with green-fluorescent protein (GFP), which was originally isolated from the cnidarian Aequorea victoria. This protein absorbs blue light (maximally at 395 nm) and emits green light (peak at 509) without the requirement of exogenous substrates and cofactors. These unique qualities allow GFP to be used to monitor gene expression and protein localization in vivo. Several mutant forms of GFP have been developed which fluoresce more intensely and have shifted excitation maxima when compared to wildtype GFP, making them useful for flow cytometry, fluorescence microscopy, and double-labeling applications. This antibody is capable of staining formaldehyde fixed cells.