Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ GFP (Green Fluorescent Protein) Ab-2, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ GFP (Green Fluorescent Protein) Ab-2, Mouse Monoclonal Antibody.

$227.92 - $1,125.35

Specifications

Antigen GFP (Green Fluorescent Protein) Ab-2
Clone GFP02 (C163)
Applications Western Blot
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1315P

 
thermo scientific™ lab vision™
MS1315P
200μg/mL 1mL Each for $1,067.80

MS1315P0

 
thermo scientific™ lab vision™
MS1315P0
200μg/mL 100μL Each for $227.92

MS1315P1

 
thermo scientific™ lab vision™
MS1315P1
200μg/mL 500μL Each for $670.08

MS1315P1ABX

 
thermo scientific™ lab vision™
MS1315P1ABX
1mg/mL 100μL Each for $751.31

MS1315PABX

 
thermo scientific™ lab vision™
MS1315PABX
1mg/mL 200μL Each for $1,125.35
Description & Specifications

Specifications

Antigen GFP (Green Fluorescent Protein) Ab-2
Clone GFP02 (C163)
Applications Western Blot
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity All
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Epitope Tags
Type Primary
Immunogen Purified recombinant GFP protein
Isotype IgG2a kappa

Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ GFP (Green Fluorescent Protein) Ab-2, Mouse Monoclonal Antibody.

GFP is a 27 kDa monomeric protein, which autocatalytically forms a fluorescent pigment. The wild type protein absorbs blue light (maximally at 395nm) and emits green light (peak emission 508nm) in the absence of additional proteins, substrates, or co-factors. GFP fluorescence is stable, species-independent and is suitable for a variety of applications. GFP has been used extensively as a fluorescent tag to monitor gene expression and protein localization. Moreover, other applications for GFP include its use in assessing protein-protein interactions in the yeast two-hybrid system, and in measuring distances between proteins in fluorescence energy transfer (FRET) experiments.

Host Species: Mouse

Clone: GFP02; same as C163

Isotype: IgG1/κ

Species Reactivity: Proteins from all species that are tagged with GFP.

Epitope: Not determined

Immunogen: Purified recombinant GFP protein

Molecular Weight: 27 kDa

Positive Control: Purified recombinant GFP protein, transfected cell lines expressing GFP-tagged proteins

Cellular Localization: Depends upon the localization of the parent protein tagged with GFP Ab-2 can be used to detect GFP and GFP variants in Western blots.

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO