Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ GFP (Green Fluorescent Protein) Ab-2, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ GFP (Green Fluorescent Protein) Ab-2, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1315PABX

 View more versions of this product

Catalog No. MS1315PABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity All
Host Species Mouse
Isotype IgG2a kappa
Immunogen Purified recombinant GFP protein
Clone GFP02 (C163)
Quantity 200µL
Applications Western Blotting
Antigen GFP (Green Fluorescent Protein) Ab-2
Regulatory Status RUO
Format Concentrated
Research Discipline Epitope Tags
Conjugate Unlabeled
Classification Monoclonal
Type Primary
Concentration 1mg/mL

Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ GFP (Green Fluorescent Protein) Ab-2, Mouse Monoclonal Antibody.

GFP is a 27 kDa monomeric protein, which autocatalytically forms a fluorescent pigment. The wild type protein absorbs blue light (maximally at 395nm) and emits green light (peak emission 508nm) in the absence of additional proteins, substrates, or co-factors. GFP fluorescence is stable, species-independent and is suitable for a variety of applications. GFP has been used extensively as a fluorescent tag to monitor gene expression and protein localization. Moreover, other applications for GFP include its use in assessing protein-protein interactions in the yeast two-hybrid system, and in measuring distances between proteins in fluorescence energy transfer (FRET) experiments.

Host Species: Mouse

Clone: GFP02; same as C163

Isotype: IgG1/κ

Species Reactivity: Proteins from all species that are tagged with GFP.

Epitope: Not determined

Immunogen: Purified recombinant GFP protein

Molecular Weight: 27 kDa

Positive Control: Purified recombinant GFP protein, transfected cell lines expressing GFP-tagged proteins

Cellular Localization: Depends upon the localization of the parent protein tagged with GFP Ab-2 can be used to detect GFP and GFP variants in Western blots.

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO