Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ GFP (Green Fluorescent Protein) Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results with Thermo Scientific™ GFP (Green Fluorescent Protein) Ab-1, Mouse Monoclonal Antibody.

$260.16 - $1,194.20

Specifications

Antigen GFP (Green Fluorescent Protein) Ab-1
Clone GFP01 (B34)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity All
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1288P

 
thermo scientific™ lab vision™
MS1288P
200μg/mL 1mL Each for $1,134.80

MS1288P0

 
thermo scientific™ lab vision™
MS1288P0
200μg/mL 100µL Each for $260.16

MS1288P1

 
thermo scientific™ lab vision™
MS1288P1
200μg/mL 500µL Each for $712.78

MS1288P1ABX

 
thermo scientific™ lab vision™
MS1288P1ABX
1mg/mL 100µL Each for $784.63

MS1288PABX

 
thermo scientific™ lab vision™
MS1288PABX
1mg/mL 200µL Each for $1,194.20
Description & Specifications

Specifications

Antigen GFP (Green Fluorescent Protein) Ab-1
Clone GFP01 (B34)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity All
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Epitope Tags
Type Primary
Immunogen Purified recombinant GFP protein
Isotype IgG2a kappa

Ensure accurate, reproducible results with Thermo Scientific™ GFP (Green Fluorescent Protein) Ab-1, Mouse Monoclonal Antibody.

GFP is a 27 kDa monomeric protein, which autocatalytically forms a fluorescent pigment. The wild type protein absorbs blue light (maximally at 395nm) and emits green light (peak emission 508nm) in the absence of additional proteins, substrates, or co-factors. GFP fluorescence is stable, species-independent and is suitable for a variety of applications. GFP has been used extensively as a fluorescent tag to monitor gene expression and protein localization. Moreover, other applications for GFP include its use in assessing protein-protein interactions in the yeast two-hybrid system, and in measuring distances between proteins in fluorescence energy transfer (FRET) experiments.

Host Species: Mouse

Clone: GFP01; same as B34

Isotype: IgG1/κ

Species Reactivity: Proteins from all species that are tagged with GFP.

Epitope: Not determined

Immunogen: Purified recombinant GFP protein

Molecular Weight: 27 kDa

Positive Control: Purified recombinant GFP protein, transfected cell lines expressing GFP-tagged proteins

Cellular Localization: Depends upon the localization of the parent protein tagged with GFP

Recommended for:

  • Western Blotting (Not verified)

Note:

USA: RUO; Int'l: RUO