Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ GFP (Green Fluorescent Protein) Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results with Thermo Scientific™ GFP (Green Fluorescent Protein) Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1288PABX

 View more versions of this product

Catalog No. MS1288PABX

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Regulatory Status RUO
Isotype IgG2a kappa
Antigen GFP (Green Fluorescent Protein) Ab-1
Cross Reactivity All
Quantity 200µL
Clone GFP01 (B34)
Immunogen Purified recombinant GFP protein
Host Species Mouse
Research Discipline Epitope Tags
Concentration 1mg/mL
Classification Monoclonal
Type Primary
Conjugate Unlabeled

Ensure accurate, reproducible results with Thermo Scientific™ GFP (Green Fluorescent Protein) Ab-1, Mouse Monoclonal Antibody.

GFP is a 27 kDa monomeric protein, which autocatalytically forms a fluorescent pigment. The wild type protein absorbs blue light (maximally at 395nm) and emits green light (peak emission 508nm) in the absence of additional proteins, substrates, or co-factors. GFP fluorescence is stable, species-independent and is suitable for a variety of applications. GFP has been used extensively as a fluorescent tag to monitor gene expression and protein localization. Moreover, other applications for GFP include its use in assessing protein-protein interactions in the yeast two-hybrid system, and in measuring distances between proteins in fluorescence energy transfer (FRET) experiments.

Host Species: Mouse

Clone: GFP01; same as B34

Isotype: IgG1/κ

Species Reactivity: Proteins from all species that are tagged with GFP.

Epitope: Not determined

Immunogen: Purified recombinant GFP protein

Molecular Weight: 27 kDa

Positive Control: Purified recombinant GFP protein, transfected cell lines expressing GFP-tagged proteins

Cellular Localization: Depends upon the localization of the parent protein tagged with GFP

Recommended for:

  • Western Blotting (Not verified)

Note:

USA: RUO; Int'l: RUO