Please login to your online account to display your discounted pricing

Alfa Aesar™ Gey's Balanced Salt Solution, with calcium, without magnesium, with phenol red

Chemical Identifiers

Chemical Name or Material Gey's Balanced Salt Solution
Name Note with calcium, without magnesium, with phenol red
MDL Number MFCD00283318
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67748AP

  SDS
alfa aesar™
J67748AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67748K2

  SDS
alfa aesar™
J67748K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Gey's Balanced Salt Solution
Name Note with calcium, without magnesium, with phenol red
Physical Form Liquid
TSCA TSCA
Quantity 500mL
Color Red
MDL Number MFCD00283318