Please login to your online account to display your discounted pricing

Alfa Aesar™ Gey's Balanced Salt Solution, with calcium, without magnesium, with phenol red


Quantity 1L

Manufacturer: alfa aesar™  J67748K2

 View more versions of this product

Catalog No. AAJ67748K2

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

MDL Number MFCD00283318
Chemical Name or Material Gey's Balanced Salt Solution
Color Red
Quantity 1L
TSCA TSCA
Name Note with calcium, without magnesium, with phenol red
Physical Form Liquid

Specifications

Chemical Name or Material Gey's Balanced Salt Solution
Name Note with calcium, without magnesium, with phenol red
MDL Number MFCD00283318