Please login to your online account to display your discounted pricing

Alfa Aesar™ Gey's Balanced Salt Solution (10X), without calcium, without magnesium, without phenol red


Quantity 500mL

Manufacturer: alfa aesar™  J67581AP

 View more versions of this product

Catalog No. AAJ67581AP

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Gey's Balanced Salt Solution (10X)
MDL Number MFCD00283318
Physical Form Liquid
Name Note without calcium, without magnesium, without phenol red
TSCA TSCA
Color Colorless
Quantity 500mL

Specifications

Chemical Name or Material Gey's Balanced Salt Solution (10X)
Name Note without calcium, without magnesium, without phenol red
MDL Number MFCD00283318