Please login to your online account to display your discounted pricing

Alfa Aesar™ Gey's Balanced Salt Solution (10X), without calcium, without magnesium, without phenol red

Chemical Identifiers

Chemical Name or Material Gey's Balanced Salt Solution (10X)
Name Note without calcium, without magnesium, without phenol red
MDL Number MFCD00283318
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67581AP

  SDS
alfa aesar™
J67581AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67581K2

  SDS
alfa aesar™
J67581K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Gey's Balanced Salt Solution (10X)
MDL Number MFCD00283318
Physical Form Liquid
Name Note without calcium, without magnesium, without phenol red
TSCA TSCA
Color Colorless
Quantity 500mL