Please login to your online account to display your discounted pricing

Alfa Aesar™ Gey's Balanced Salt Solution (10X), with calcium, with magnesium, with phenol red


Quantity 500mL

Manufacturer: alfa aesar™  J67602AP

 View more versions of this product

Catalog No. AAJ67602AP

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

MDL Number MFCD00283318
Physical Form Liquid
TSCA TSCA
Color Red
Quantity 500mL
Chemical Name or Material Gey's Balanced Salt Solution (10X)
Name Note with calcium, with magnesium, with phenol red

Specifications

Chemical Name or Material Gey's Balanced Salt Solution (10X)
Name Note with calcium, with magnesium, with phenol red
MDL Number MFCD00283318