Please login to your online account to display your discounted pricing

Alfa Aesar™ Gey's Balanced Salt Solution (10X), with calcium, with magnesium, with phenol red

Chemical Identifiers

Chemical Name or Material Gey's Balanced Salt Solution (10X)
Name Note with calcium, with magnesium, with phenol red
MDL Number MFCD00283318
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67602AP

  SDS
alfa aesar™
J67602AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67602K2

  SDS
alfa aesar™
J67602K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

MDL Number MFCD00283318
Physical Form Liquid
TSCA TSCA
Color Red
Quantity 500mL
Chemical Name or Material Gey's Balanced Salt Solution (10X)
Name Note with calcium, with magnesium, with phenol red