Please login to your online account to display your discounted pricing

Alfa Aesar™ Gey's Balanced Salt Solution (10X), with calcium, with magnesium, with phenol red


Quantity 1L

Manufacturer: alfa aesar™  J67602K2

 View more versions of this product

Catalog No. AAJ67602K2

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Gey's Balanced Salt Solution (10X)
MDL Number MFCD00283318
Physical Form Liquid
TSCA TSCA
Color Red
Quantity 1L
Name Note with calcium, with magnesium, with phenol red

Specifications

Chemical Name or Material Gey's Balanced Salt Solution (10X)
Name Note with calcium, with magnesium, with phenol red
MDL Number MFCD00283318