Learn More
Germanium tetrachloride, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 197150250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisSpecifications
82.0°C to 84.0°C | |
-49.5°C | |
10038-98-9 | |
Liquid | |
99.99% | |
GeCl4 | |
15, 4444 | |
Solubility in water: decomposes. Other solubilities: soluble in benzene, ether, carbon disulfid and, other organic solvents, insoluble in concentrated hydrochloric acid | |
Cl.Cl.Cl.Cl.[GeH4] | |
222.49 | |
214.4 | |
Trace Metal Basis | |
1.84 |
Germanium tetrachloride | |
99.99% | |
Colorless | |
25 g | |
Cl4GeH8 | |
MFCD00011029 | |
germanium tetrachloride, germanium chloride, germane, tetrachloro, germanium iv chloride, germanium, tetrachloro, gecl4, germanium chloride gecl4, unii-ysv1r803c0, wln: .ge..g4, germanium chloride gecl4 8ci | |
VDHBHZZBEGQJRK-UHFFFAOYSA-N | |
tetrachlorogermane | |
66226 | |
99.99% | |
Glass bottle | |
1.8400g/mL |
Chemical Identifiers
10038-98-9 | |
222.49 | |
VDHBHZZBEGQJRK-UHFFFAOYSA-N | |
66226 |
Cl4GeH8 | |
MFCD00011029 | |
germanium tetrachloride, germanium chloride, germane, tetrachloro, germanium iv chloride, germanium, tetrachloro, gecl4, germanium chloride gecl4, unii-ysv1r803c0, wln: .ge..g4, germanium chloride gecl4 8ci | |
Cl.Cl.Cl.Cl.[GeH4] |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.
May cause respiratory irritation.
Contact with water liberates toxic gas.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 233-116-7
RTECSNumber : LY5225000
TSCA : TSCA
RUO – Research Use Only