missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Germanium powder, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1019018

 View more versions of this product

Catalog No. AA1019018



Specifications

Specifications

Germanium
99.9999% (Metals basis)
MFCD00085310
atom
[Ge]
72.63
CHEBI:30441
Odorless
Silver
7440-56-4
Ge
Silver-colored powder
GNPVGFCGXDBREM-UHFFFAOYSA-N
germanium
6326954
Powder
0 hPa
50g

Chemical Identifiers

7440-56-4
72.63
GNPVGFCGXDBREM-UHFFFAOYSA-N
6326954
germanium
Ge
MFCD00085310
atom
CHEBI:30441
[Ge]
Safety and Handling

Safety and Handling

EINECSNumber : 231-164-3

Documents