missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Germanium, Plasma standard solution, Ge 1000μg/mL, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13841AP

 View more versions of this product

Catalog No. AA13841AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Germanium
Liquid
MFCD00085310
GNPVGFCGXDBREM-UHFFFAOYSA-N
germanium
6326954
Liquid
500mL
7440-56-4,7664-39-3,7697-37-2,7732-18-5
Ge
Nitric acid: 5.0%; Hydrogen fluoride: 0.1%; Germanium: 0.1%; Water: 94.8%
[Ge]
72.63
CHEBI:30441
23 hPa (17mm Hg) at 20°C

Chemical Identifiers

7440-56-4
72.63
GNPVGFCGXDBREM-UHFFFAOYSA-N
CHEBI:30441
[Ge]
Ge
MFCD00085310
6326954
germanium
Safety and Handling

Safety and Handling

H314-H303-H313-H333: Causes severe skin burns and eye damage. May be harmful if swallowed. May be harmful in contact with skin. May be harmful if inhaled.

DOTInformation : Hazard Class: 8, Packing Group: III

Documents