missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Germanium pieces, 99.999% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4398609

 View more versions of this product

Catalog No. AA4398609



Specifications

Specifications

Germanium
99.9999% (Metals basis)
MFCD00085310
element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
[Ge]
72.63
CHEBI:30441
5.32g/cm3
0 hPa
10g
7440-56-4
Ge
Silver-colored pieces
GNPVGFCGXDBREM-UHFFFAOYSA-N
germanium
6326954
Pieces, 3.2mm (0.125 in.) and down
Odorless
Silver

Chemical Identifiers

7440-56-4
72.63
GNPVGFCGXDBREM-UHFFFAOYSA-N
6326954
germanium
Ge
MFCD00085310
element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
CHEBI:30441
[Ge]
Safety and Handling

Safety and Handling

EINECSNumber : 231-164-3

Documents