missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Germanium(IV) oxide, White powder/flake/crystalline/beads, etc., 99.9999% (Metals basis), Alfa Aesar™

$97.08 - $1,092.89

Chemical Identifiers

CAS 1310-53-8
Molecular Formula GeO2
Molecular Weight (g/mol) 104.628
MDL Number MFCD00011030
InChI Key YBMRDBCBODYGJE-UHFFFAOYSA-N
Synonym dioxogermanium, germania, germanic acid, germanic oxide, germanic oxide crystalline, germanium dioxide, germanium iv oxide, germanium oxide, wln: ge o2
PubChem CID 14796
IUPAC Name dioxogermane
SMILES O=[Ge]=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1051006
View Documents
Thermo Fisher Scientific
1051006
5g
Each for $97.08
 
AA1051014
View Documents
Thermo Fisher Scientific
1051014
25g
Each for $371.26
 
AA1051022
View Documents
Thermo Fisher Scientific
1051022
100g
Each for $1,092.89
 
Description

Description

Specifications

Chemical Identifiers

1310-53-8
104.628
YBMRDBCBODYGJE-UHFFFAOYSA-N
14796
O=[Ge]=O
GeO2
MFCD00011030
dioxogermanium, germania, germanic acid, germanic oxide, germanic oxide crystalline, germanium dioxide, germanium iv oxide, germanium oxide, wln: ge o2
dioxogermane

Specifications

1310-53-8
GeO2
dioxogermanium, germania, germanic acid, germanic oxide, germanic oxide crystalline, germanium dioxide, germanium iv oxide, germanium oxide, wln: ge o2
YBMRDBCBODYGJE-UHFFFAOYSA-N
dioxogermane
14796
-40 Mesh Powder
Odorless
1086°C
Germanium(IV) oxide
99.9999% (Metals basis)
MFCD00011030
Soluble in water (4.5g/L at 25°C),acid and alkali. Insoluble in Hydrofluoric acid and hydrochloric acid.
O=[Ge]=O
104.628
104.59
6.239
White
5g
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

EINECSNumber : 215-180-8

RTECSNumber : LY5240000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS