Learn More
Alfa Aesar™ Germanium(IV) iodide, 99.999% (metals basis)
Manufacturer: Alfa Aesar™ 1400918
Specifications
Germanium(IV) iodide | |
(metals basis) | |
MFCD00016120 | |
germanium tetraiodide, germane, tetraiodo, germanium iv iodide, germanium iodide, acmc-1bq9p, germanium iv iodide trace metals basis | |
[Ge](I)(I)(I)I | |
580.248 | |
580.21 | |
99.999% | |
−10 Mesh | |
146°C |
13450-95-8 | |
GeI4 | |
UN3260 | |
CUDGTZJYMWAJFV-UHFFFAOYSA-N | |
tetraiodogermane | |
83479 | |
Crystalline | |
4.32g/mL | |
Moisture Sensitive | |
50g |
Chemical Identifiers
13450-95-8 | |
580.248 | |
CUDGTZJYMWAJFV-UHFFFAOYSA-N | |
83479 | |
[Ge](I)(I)(I)I |
GeI4 | |
MFCD00016120 | |
germanium tetraiodide, germane, tetraiodo, germanium iv iodide, germanium iodide, acmc-1bq9p, germanium iv iodide trace metals basis | |
tetraiodogermane |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
TSCA : TSCA