missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Germanium(IV) ethoxide, 99.995% (metals basis)

Preparation of GeO2 by CVD; intermediate in sol-gel synthesis

Chemical Identifiers

CAS 14165-55-0
Molecular Formula C8H20GeO4
Linear Formula Ge(OC2H5)4
Formula Weight 252.84
MDL Number MFCD00015121
UN Number UN1993
Synonym Germanium tetraethoxide; Tetraethoxygermane
Chemical Name or Material Germanium (IV) ethoxide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4305203 SDS Alfa Aesar™
4305203
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA4305214 SDS Alfa Aesar™
4305214
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA4305206 SDS Alfa Aesar™
4305206
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

14165-55-0
Ge(OC2H5)4
MFCD00015121
Germanium tetraethoxide; Tetraethoxygermane
C8H20GeO4
252.84
UN1993
Germanium (IV) ethoxide

Specifications

185.5°C
14165-55-0
49°C (121°F)
Colorless
1.4070
Liquid
Ge(OC2H5)4
MFCD00015121
Germanium tetraethoxide; Tetraethoxygermane
99.995%, (metals basis)
−72°C
1g
1.14g/mL
Alcohol-like
Moisture Sensitive
C8H20GeO4
252.84
UN1993
Germanium (IV) ethoxide

Safety and Handling

GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280g-P305+P351+P338-P337+P313
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA