missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Germanium(IV) ethoxide, 97%

Manufacturer:  Alfa Aesar™ 1467903

 View more versions of this product

Catalog No. AA1467903



Specifications

Specifications

Germanium(IV) ethoxide
-72°C
1g
1.14g/mL
Alcohol-like
Moisture Sensitive
MFCD00015121
Germanium tetraethoxide; Tetraethoxygermane
252.84
97%
185.5°C
14165-55-0
49° (121°F)
Colorless
1.4070
Ge(OC2H5)4
UN1993
Soluble in anhydrous alcohol and benzene; Hydrolyzes in water
Liquid

Chemical Identifiers

14165-55-0
MFCD00015121
Germanium tetraethoxide; Tetraethoxygermane
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280g-P305+P351+P338-P337+P313
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

SDS Documents