missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Germanium(IV) chloride, 99.9999% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 1050922

 View more versions of this product

Catalog No. AA1050922



Description

Description

It is used as a catalyst in the conversion of carbohydrates into 5-hydroxymethylfurfural in ionic liquids, as a reducing agent in combination with triphenylphosphine (TPP) for the reduction of alpha-bromo carboxylic acid derivatives, and in the production of pure germanium. It also finds use as an intermediate for several optical processes. It is one of the most important dopants in silica glass for optical fibers. It is employed in the preparation of germanium dioxide which is used for wide camera lens, microscopy, IR-transparent glasses/windows/lenses, and for the core of fiber-optic lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

83.1°C
-49.5°C
Colorless
100 g
Cl4GeH8
MFCD00011029
14,4410
Soluble in ether,benzene,chloroform,carbon tetrachloride. Insoluble in hydrochloric acid and sulphuric acid.
Cl.Cl.Cl.Cl.[GeH4]
222.49
214.4
Acidic
1.874 g/mL
Germanium(IV) chloride
10038-98-9
Liquid
(metals basis)
GeCl4
UN3264
germanium tetrachloride, germanium chloride, germane, tetrachloro, germanium iv chloride, germanium, tetrachloro, gecl4, germanium chloride gecl4, unii-ysv1r803c0, wln: .ge..g4, germanium chloride gecl4 8ci
VDHBHZZBEGQJRK-UHFFFAOYSA-N
germanium tetrachloride
66226
99.9999%
Moisture sensitive

Chemical Identifiers

10038-98-9
222.49
VDHBHZZBEGQJRK-UHFFFAOYSA-N
66226
Cl4GeH8
MFCD00011029
germanium tetrachloride, germanium chloride, germane, tetrachloro, germanium iv chloride, germanium, tetrachloro, gecl4, germanium chloride gecl4, unii-ysv1r803c0, wln: .ge..g4, germanium chloride gecl4 8ci
Cl.Cl.Cl.Cl.[GeH4]
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: I

EINECSNumber : 233-116-7

RTECSNumber : LY5225000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only