missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Germanium Ingot, zone refined, 95% N type min., 99.9999% (Metals basis, 95% Ntype min.), Alfa Aesar™

Form: Electrical Resistivity: 40 ohm-cm

Manufacturer:  Alfa Aesar 10192TK

 View more versions of this product

Catalog No. AA10192TK



Specifications

Specifications

Germanium
99.9999% (Metals basis, 95% N type minimum)
MFCD00085310
element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
[Ge]
72.63
CHEBI:30441
Odorless
24g
7440-56-4
Ge
Silver-colored ingot
GNPVGFCGXDBREM-UHFFFAOYSA-N
germanium
6326954
Ingot, Zone Refined, 95% N Type Minimum
Silver

Chemical Identifiers

7440-56-4
72.63
GNPVGFCGXDBREM-UHFFFAOYSA-N
6326954
germanium
Ge
MFCD00085310
element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
CHEBI:30441
[Ge]
Documents