Please login to your online account to display your discounted pricing

Alfa Aesar™ Germanium(II) telluride, 99.999%

$77.46 - $268.20

Chemical Identifiers

Chemical Name or Material Germanium(II) telluride
CAS 12025-39-7
Molecular Formula GeTe
Formula Weight 200.19
MDL Number MFCD00135541
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2112803

  SDS
alfa aesar™
B2112803
1g Each for $77.46

AAB2112806

  SDS
alfa aesar™
B2112806
5g Each for $268.20
Description & Specifications

Specifications

Formula Weight 200.19
Odor Odorless
TSCA TSCA
Quantity 1g
MDL Number MFCD00135541
Health Hazard 1 Warning
Molecular Formula GeTe
Assay 99.999%
CAS 12025-39-7
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Melting Point 725°C
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Germanium(II) telluride

Applications
It is a promising thermoelectric compound with uses in doping, alloying, and nanostructuring, non-stoichiometric defects. It is also an important semiconductor that can act as a low temperature superconductor when doped. GeTe is also an important semiconductor material prototypical phase change material of high interest for applications in optical and electronic non-volatile memories.

Solubility
Insoluble in water.

Notes
Store away from acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.