missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Germanium(II) iodide, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 1401303

 View more versions of this product

Catalog No. AA1401303



Specifications

Specifications

Germanium(II) iodide
(metals basis)
MFCD00135537
germanium diiodide, germanium ii iodide, germanium iodide, diiodogermanylidene, acmc-1c83a, germanium ii iodide trace metals basis
[Ge](I)I
326.439
326.4
5.37g/mL
Air & Moisture Sensitive
1g
13573-08-5
GeI2
UN3260
IAGYEMVJHPEPGE-UHFFFAOYSA-N
diiodogermanium
6327215
99.99%
−10 Mesh
Yellow to Brown

Chemical Identifiers

13573-08-5
326.439
IAGYEMVJHPEPGE-UHFFFAOYSA-N
6327215
[Ge](I)I
GeI2
MFCD00135537
germanium diiodide, germanium ii iodide, germanium iodide, diiodogermanylidene, acmc-1c83a, germanium ii iodide trace metals basis
diiodogermanium
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Documents