Learn More
Alfa Aesar™ Germanium(II) iodide, 99.99% (metals basis)
Manufacturer: Alfa Aesar™ 1401306
Specifications
Germanium(II) iodide | |
(metals basis) | |
MFCD00135537 | |
germanium diiodide, germanium ii iodide, germanium iodide, diiodogermanylidene, acmc-1c83a, germanium ii iodide trace metals basis | |
[Ge](I)I | |
326.439 | |
326.4 | |
5.37g/mL | |
Air & Moisture Sensitive | |
5g |
13573-08-5 | |
GeI2 | |
UN3260 | |
IAGYEMVJHPEPGE-UHFFFAOYSA-N | |
diiodogermanium | |
6327215 | |
99.99% | |
−10 Mesh | |
Yellow to Brown |
Chemical Identifiers
13573-08-5 | |
326.439 | |
IAGYEMVJHPEPGE-UHFFFAOYSA-N | |
6327215 | |
[Ge](I)I |
GeI2 | |
MFCD00135537 | |
germanium diiodide, germanium ii iodide, germanium iodide, diiodogermanylidene, acmc-1c83a, germanium ii iodide trace metals basis | |
diiodogermanium |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
TSCA : TSCA