missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Germanium, 99.999%, (trace metal basis), powder, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 193110100

 View more versions of this product

Catalog No. AC193110100



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Specifications

2830.0°C
937.0°C
7440-56-4
Glistening Crystalline Powder
99.999%
MFCD00085310
element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
GNPVGFCGXDBREM-UHFFFAOYSA-N
germanium
6326954
72.59
Trace Metal Basis
5.35
Germanium
99.999%
Black to Gray
10 g
Ge
15, 4441
Solubility in water: insoluble. Other solubilities: insoluble in hcl and diluted alkali hydroxides
[Ge]
72.63
CHEBI:30441
99.999%
Glass bottle
5.3500g/mL

Chemical Identifiers

7440-56-4
72.63
GNPVGFCGXDBREM-UHFFFAOYSA-N
6326954
germanium
Ge
MFCD00085310
element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
CHEBI:30441
[Ge]
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 231-164-3

RTECSNumber : LY5200000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only