Please login to your online account to display your discounted pricing

Vileda Professional™ Geo Metallic Waste Management Bins

Designed to meet the challenge contemporary buildings and interiors place upon the products used inside them.

$147.72 - $176.90

Specifications

Material
  • Bin Body: 100% Polypropylene
  • Rubber Feet: 100% Thermoplastic elastomer
  • Clips:100% Polypropylene
Finish Metallic
For Use With (Application) Ideal for ISO 7, LAR, Production and Labs
Shape Rectangular
Color Metallic/Black
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Product Type Price Quantity    

19-200-721

 
vileda professional™
137660
14.5 gal Medium bin Each for $147.72
Case of 4 Each for $554.56

19-200-722

 
vileda professional™
137661
18.5 gal. Large bin Each for $176.90
Case of 4 Each for $664.08
Description & Specifications

Specifications

Material
  • Bin Body: 100% Polypropylene
  • Rubber Feet: 100% Thermoplastic elastomer
  • Clips:100% Polypropylene
Finish Metallic
For Use With (Application) Ideal for ISO 7, LAR, Production and Labs
Shape Rectangular
Color Metallic/Black

By utilizing the “combined materials” technology, merging the durability and performance of plastic with the aesthetics and image of stainless steel. The result is a bin that is easy to clean, fingerprint, rust and dent proof - plus looks at home in even the latest state-of-the-art interiors.

  • Durable plastic construction with metallic finish
  • Compact slim line design – fits in the tightest spaces
  • Rust, dent and fingerprint proof
  • Easy to clean and washable
  • Integrated handle - easy carrying and lifting
  • Integrated bag holder – ensures the correct fit of your waste bag
  • Anti-slip rubber feet – avoids movement in use and protects flooring