Please login to your online account to display your discounted pricing

Gentian Violet, Reagent Grade, LabChem™

$86.40 - $86.40

Chemical Identifiers

Chemical Name or Material Gentian Violet
CAS 548-62-9
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula C25H30CIN3
Formula Weight 407.99
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC147908

 
lab chem
LC147908
Amber Glass 25g Each for $86.40
Description & Specifications

Specifications

Formula Weight 407.99
Chemical Name or Material Gentian Violet
Quantity 25g
EINECS Number 208-953-6
Purity Grade Reagent Grade
Physical Form Powder
Linear Formula C25H30CIN3
CAS 548-62-9
Identification Passes Test
Health Hazard 1 Danger
Packaging Amber Glass
CAS Max % 100
Health Hazard 2 GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Suspected of causing cancer.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not eat, drink or smoke when using this product.
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse month.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Solubility Soluble in water
Specific Gravity 1.19g/cm3
Recommended Storage Room Temperature
Decomposition Information Carbon monoxide; Carbon dioxide; Nitrogen oxides
Density 1.19g/cm3
Melting Point 173°C
Color Greenish
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not eat, drink or smoke when using this product.
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse month.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Suspected of causing cancer.
Very toxic to aquatic life with long lasting effects.

Room Temperature