missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gentian Violet, Hucker Formulation, for Gram Staining, Ricca Chemical

Chemical Identifiers

CAS 7732-18-5,64-17-5,548-62-9,6009-70-7,7732-18-5 (Water, Deionized)
CAS Min % 82.0,13.2,1.65,0.66
CAS Max % 86.0,13.74,1.72,0.69
Physical Form Liquid
Chemical Name or Material Gentian Violet, Hucker Formulation
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
3240-1 Ricca Chemical Company
RI32401
Poly natural 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
3240-4 Ricca Chemical Company
RI32404
Poly natural 120mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
3240-32 SDS Ricca Chemical Company
RI324032
Poly natural 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Gram Staining
Description & Specifications

Chemical Identifiers

7732-18-5,64-17-5,548-62-9,6009-70-7,7732-18-5 (Water, Deionized)
86.0,13.74,1.72,0.69
Gentian Violet, Hucker Formulation
82.0,13.2,1.65,0.66
Liquid

Specifications

7732-18-5,64-17-5,548-62-9,6009-70-7,7732-18-5 (Water, Deionized)
86.0,13.74,1.72,0.69
Poly natural
4L
82.0,13.2,1.65,0.66
Liquid
Clear dark blue-purple
Gentian Violet, Hucker Formulation