Please login to your online account to display your discounted pricing

Gentian Violet, 2% Methanolic, Certified, LabChem™

$37.91 - $37.91

Chemical Identifiers

Chemical Name or Material Gentian Violet
CAS 67-56-1
CAS Max % 98
CAS 548-62-9
CAS Max % 2
Name Note 2% Methanolic
Purity Grade Certified Grade
Linear Formula C25H30CIN3
Formula Weight 407.99
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC148307

 
lab chem
LC148307
Poly Bottle 125mL Each for $37.91
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 407.99
Health Hazard 1 Danger
Physical Form Liquid
CAS Max % 98
Packaging Poly Bottle
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide; Nitrogen oxides
UN Number UN1993
Solubility Soluble in water
CAS Max % 2
Recommended Storage Room Temperature
Quantity 125mL
Linear Formula C25H30CIN3
Color Dark purple
Identification Passes Test
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed, in contact with skin or if inhaled.
Causes serious eye irritation.
Suspected of causing cancer.
Causes damage to organs (central nervous system, optic nerve).
Harmful to aquatic life.
Harmful to aquatic life with long lasting effects.
CAS 548-62-9
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Cal a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use alcohol resistant foam, extinguishing powder to extinguish.
Store locked up in a cool well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Chemical Name or Material Gentian Violet
Name Note 2% Methanolic
CAS 67-56-1
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Cal a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use alcohol resistant foam, extinguishing powder to extinguish.
Store locked up in a cool well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed, in contact with skin or if inhaled.
Causes serious eye irritation.
Suspected of causing cancer.
Causes damage to organs (central nervous system, optic nerve).
Harmful to aquatic life.
Harmful to aquatic life with long lasting effects.

Room Temperature