missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gentian Violet, 1% Alcoholic, Certified, LabChem™

$62.46 - $62.46

Chemical Identifiers

CAS 64-17-5,67-63-0,67-56-1,548-62-9
CAS Max % 89.1,4.95,4.95,1
Linear Formula C25H30CIN3
Formula Weight 407.99
UN Number UN1170
Chemical Name or Material Gentian Violet, 1% Alcoholic
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
LC148001 Lab Chem
LC148001
Poly Bottle 500mL Each for $62.46
Specifications Description & Specifications

Chemical Identifiers

64-17-5,67-63-0,67-56-1,548-62-9
C25H30CIN3
UN1170
Certified Grade
89.1,4.95,4.95,1
407.99
Gentian Violet, 1% Alcoholic

Specifications

64-17-5,67-63-0,67-56-1,548-62-9
Liquid
407.99
Passes Test
Soluble in water
Fume; Carbon monoxide; Carbon dioxide; May release flammable gases
500mL
Certified Grade
89.1,4.95,4.95,1
C25H30CIN3
UN1170
Poly Bottle
Passes Test
Dark purple
Gentian Violet, 1% Alcoholic

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve).

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe vapors, mist, spray.
Do not eat, drink or smoke when using this product.
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse month.
Call a poison center/doctor if you feel unwell.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature