missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gentamicin sulfate solution-10, MP Biomedicals™
Manufacturer: Mp Biomedicals 0210503080
Specifications
Gentamycin Sulfate Solution - 10 | |
1405-41-0 | |
Garamycin, Genoptic | |
USP | |
100mL |
Cell Biology and Analysis | |
MFCD00270181 | |
Liquid | |
Colorless to Yellow |
Chemical Identifiers
1405-41-0 | |
MFCD00270181 | |
Garamycin, Genoptic |
Safety and Handling
EINECSNumber : 215-778-9
Recommended Storage : Store at 4°C.