missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals Inc Gentamicin sulfate solution-10, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210503050
Specifications
Gentamycin Sulfate | |
Cell Biology and Analysis | |
1405-41-0 | |
Garamycin, Genoptic | |
Colorless to Yellow |
Gentamycin Sulfate Solution - 10 | |
Liquid | |
MFCD00270181 | |
USP | |
5 x 10 mL |
Chemical Identifiers
1405-41-0 | |
MFCD00270181 | |
Garamycin, Genoptic |
Safety and Handling
EINECSNumber : 215-778-9
Recommended Storage : Store at 4°C.