missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gentamicin sulfate solution-10, MP Biomedicals™

$68.53 - $311.01

Chemical Identifiers

CAS 1405-41-0
MDL Number MFCD00270181
Synonym Garamycin, Genoptic
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10503010
View Documents
Mp Biomedicals
0210503010
10mL
Each for $68.53
 
ICN10503050
View Documents
Mp Biomedicals
0210503050
5 x 10mL
Each for $156.86
 
ICN10503080
View Documents
Mp Biomedicals
0210503080
100mL
Each for $311.01
 
Description

Description

Specifications

Chemical Identifiers

1405-41-0
MFCD00270181
Garamycin, Genoptic

Specifications

1405-41-0
Garamycin, Genoptic
USP
10mL
MFCD00270181
Liquid
Colorless to Yellow
Gentamycin Sulfate Solution - 10, Cell Biology and Analysis
Safety and Handling

Safety and Handling

EINECSNumber : 215-778-9

Recommended Storage : Store at 4°C.

SDS
Product Certifications